792373
2-Chloro-2,2-difluoroacetophenone
95%
Synonym(s):
2-Chloro-2,2-difluoro-1-phenylethanone, Chlorodifluoromethyl phenyl ketone, α-Chloro-α,α-difluoroacetophenone
About This Item
Recommended Products
Assay
95%
form
liquid
reaction suitability
reaction type: C-C Bond Formation
refractive index
n20/D 1.4954
n20/D 1.497 (lit.)
bp
94-96 °C/35 mmHg (lit.)
density
1.293 g/mL at 25 °C (lit.)
1.3178 g/mL at 25 °C
functional group
chloro
fluoro
ketone
phenyl
SMILES string
FC(F)(Cl)C(=O)c1ccccc1
InChI
1S/C8H5ClF2O/c9-8(10,11)7(12)6-4-2-1-3-5-6/h1-5H
InChI key
MNOONJNILVDLSW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- As a reagent in the difluoromethylation of various phenols to yield aryl difluoromethyl ethers.
- As a precursor in the Baylis-Hillman reaction of fluoroalkyl ketones to obtain chlorodifluoromethyl containing products.
- As a substrate in the synthesis of propargyl alcohols using a novel ruthenium catalyst.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
160.0 °F
Flash Point(C)
71.1 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service