Skip to Content
Merck
All Photos(1)

Documents

791997

Sigma-Aldrich

Vanadyl phthalocyanine

dye content >90 %

Synonym(s):

Oxo[29H,31H-phthalocyaninato(2-)-κN29,κN30,κN31,κN32]vanadium, Vanadium(IV)phthalocyanine oxide, Xerox xpp-VOPc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16N8OV
CAS Number:
Molecular Weight:
579.46
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder (blue)

composition

dye content, >90%

λmax

701 nm

SMILES string

O=[V]1N(/C(C2=C/3C=CC=C2)=N\C4=N/C(C5=C4C=CC=C5)=N\6)C3=N/C(C7=C/8C=CC=C7)=NC8=N/C9=C%10C(C=CC=C%10)=C6N91

InChI

1S/C32H16N8.O.V/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;;/q-2;;+2

InChI key

YRZZLAGRKZIJJI-UHFFFAOYSA-N

Legal Information

Product of Xerox®.
Xerox is a registered trademark of Xerox Corporation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service