Skip to Content
Merck
All Photos(1)

Key Documents

779776

Sigma-Aldrich

Methyl-β-cyclodextrin

produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% cyclodextrin basis (calculated)

Synonym(s):

Cavasol® W7 M, Methyl-beta-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

Assay

≥95.0% cyclodextrin basis (calculated)

form

powder

extent of labeling

1.6-1.9 molar substitution

impurities

≤0.2% unsubstituted cyclodextrin
≤0.2% volatile organics
≤2% chloride

loss

≤7.0% loss on drying

mp

180-182 °C (lit.)

SMILES string

O1C2OC(C(OC3OC(C(OC4OC(C(OC5OC(C(OC6OC(C(OC7OC(C(OC8OC(C1C(C8OC)O)COC)C(C7OC)O)COC)C(C6OC)O)COC)C(C5OC)O)COC)C(C4OC)O)COC)C(C3OC)O)COC)C(C2OC)O)COC

InChI

1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3

InChI key

QGKBSGBYSPTPKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl-β-cyclodextrin (M-β-CD) can be used as a reactant to prepare ternary inclusion complex of diclofenac (DCF) in the presence of monoethanolamine (MEA). This complex improves the percutaneous absorption of DCF in human skin. It can also be used to form an inclusion complex with olanzapine (OLP) to increase its solubility for efficient drug delivery.

Other Notes

For R&D Use

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

368.6 °F

Flash Point(C)

187 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Julia Mora et al.
European journal of medicinal chemistry, 45(9), 4079-4088 (2010-06-23)
Here, we describe the chemical characterization of the inclusion complex between diclofenac (DCF) and methyl-beta-cyclodextrin (M-beta-CD) in the presence or absence of monoethanolamine (MEA). Several techniques were used to analyze the complex both in solution and in the solid state.
Inclusion complex of methyl-β -cyclodextrin and olanzapine as potential drug delivery system for schizophrenia
de Freitas MR, et al.
Carbohydrate Polymers, 89(4), 1095-1100 (2012)
Mario Menschikowski et al.
Cancer cell international, 11, 4-4 (2011-02-16)
Increasing evidences show that beyond its role in coagulation, endothelial protein C receptor (EPCR) interferes with carcinogenesis. Pro-carcinogenic effects of EPCR were linked with a raised generation of activated protein C (aPC) and anti-apoptotic signalling. This study was carried out
William E Brownell et al.
Pflugers Archiv : European journal of physiology, 461(6), 677-686 (2011-03-05)
Changing the concentration of cholesterol in the plasma membrane of isolated outer hair cells modulates electromotility and prestin-associated charge movement, suggesting that a similar manipulation would alter cochlear mechanics. We examined cochlear function before and after depletion of membrane cholesterol
Laurie Hamtiaux et al.
PloS one, 6(10), e26823-e26823 (2011-11-03)
The antitumoral properties of endocannabinoids received a particular attention these last few years. Indeed, these endogenous molecules have been reported to exert cytostatic, apoptotic and antiangiogenic effects in different tumor cell lines and tumor xenografts. Therefore, we investigated the cytotoxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service