Skip to Content
Merck
All Photos(1)

Key Documents

779253

Sigma-Aldrich

1-Azido-4-fluorobenzene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

4-Fluorophenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4FN3
CAS Number:
Molecular Weight:
137.11
Beilstein:
1939307
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥95.0% (HPLC)

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C6H4FN3/c7-5-1-3-6(4-2-5)9-10-8/h1-4H

InChI key

VHFLTICYUZLEII-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decomposition of halogen-substituted phenyl azides
Smalley, R. K. and Suschitzky, H.
Journal of the Chemical Society, 5922-5925 (1964)
Mingyu Hu et al.
Organic letters, 10(24), 5529-5531 (2008-12-05)
A panel of small molecule-based MMP inhibitors containing rhodanine warheads was assembled using "one-pot" click chemistry. Upon biological screening, moderate inhibitors were identified which specifically targets MMP-7 and MMP-13 over other MMPs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service