Skip to Content
Merck
All Photos(1)

Key Documents

771937

Sigma-Aldrich

Platinum

nanopowder, 200 nm particle size (SEM), 99.9% (metals basis)

Synonym(s):

Platinum black, Platinum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% (metals basis)

form

nanopowder

reaction suitability

reagent type: catalyst
core: platinum

resistivity

10.6 μΩ-cm, 20°C

particle size

200 nm (SEM)

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

Platinum nanopowder has a high catalytic activity and is used widely in catalysis, batteries, fuel cells, capacitors and sensors. They are used in the synthesis of counter electrode in dyes sensitized solar cells. Since it is difficult to directly use platinum nanoparticle as the electrode, they are usually surface supported on graphene or multi-walled carbon nanotubes.

Application

Nanoscale platinum powders possess improved reaction rates compared to bulk powders owing to their high surface area. Platinum nanopowders have found use in applications including in fuel cells and oxidation catalysts.

Features and Benefits

High catalytic functionality, high conductivity and chemical stability in an acid-base environment.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Electrodeposited platinum nanoparticles on the multi-walled carbon nanotubes and its electrocatalytic for nitric oxide
Zhang, L., Fang, Z., Zhao, G. C., & Wei, X. W.
International Journal of Electrochemical Science, 3(6), 746-754 (2008)
A Biologically-Inspired Electrochemical Half-Cell for Light-Driven Generation of Hydrogen
Rebecca A. Grimme and John H. Golbeck
Material Matters, 3(4) (2008)
Yu;X.;Ye;S.;
Journal of Power Sources, 172(1), 145-154 (2007)
Keith B Male et al.
The Analyst, 132(12), 1254-1261 (2008-03-06)
Platinum nanoparticles were electrodeposited by a multi-potential step technique onto a multi-walled carbon nanotube (MWCNT) film pre-casted on a glassy carbon (GC) or boron-doped diamond (BDD) electrode. The MWCNT network consisted of Pt nanoparticles with an average diameter of 120
Fabrication of platinum nanoparticle counter electrode for highly efficient dye-sensitized solar cells by controlled thermal reduction time
Hoa, N. T. Q., Dao, V. D., & Choi, H. S.
J. Mater. Sci., 49(14), 4973-4978 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service