Skip to Content
Merck
All Photos(1)

Key Documents

767905

Sigma-Aldrich

4,4′-Dimethoxydiphenylamine

99%

Synonym(s):

4-Biphenylamine,4,4′-dimethoxy-, 4-Methoxy-N-(4-methoxyphenyl)aniline, p,p′-Dimethoxydiphenylamine, Benzenamine,4-methoxy-N-(4-methoxypheny)-, Bis(4-methoxyphenyl)-amine, Bis(4-methoxyphenyl)amine, Bis(p-anisyl)amine, Bis(p-methoxyphenyl)amine, Di-p-anisylamine, Di-p-methoxyphenylamine, Termofleks A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15NO2
CAS Number:
Molecular Weight:
229.27
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

mp

100-104 °C

SMILES string

COc1ccc(Nc2ccc(OC)cc2)cc1

InChI

1S/C14H15NO2/c1-16-13-7-3-11(4-8-13)15-12-5-9-14(17-2)10-6-12/h3-10,15H,1-2H3

InChI key

VCOONNWIINSFBA-UHFFFAOYSA-N

General description

4,4′-Dimethoxydiphenylamine is an aromatic amine that is used as a hole transporting material that facilitates efficiency mobility of charges in an electrochemical device.

Application

It can be used as a hole transporting material in the synthesis of 4,4′-dimethoxydiphenylamine-substituted 9,9′- bifluorenylidene with a power efficiency of ~18% which may be used in combination with an electron transporting layer in the fabrication of polymeric solar cells. It may also be used in the preparation of dimethyldiphenylamino functionalized carbazoles as electronically active materials which can potentially be used in the development of optoelectronic devices.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diphenylamino-substituted derivatives of 9-phenylcarbazole as glass-forming hole-transporting materials for solid state dye sensitized solar cells.
Tomkeviciene A, et al.
Synthetic Metals, 162(23), 1997-2004 (2012)
Triarylene linked spacer effect for dye-sensitized solar cells
Chang, Y. J., Wu, Y. J., Chou, P. T., Watanabe, M., & Chow, T. J.
Thin Solid Films, 558, 330-336 (2014)
Thermally stable triaryl amino chromophores with high molecular hyperpolarizabilities
Spraul, Bryan K., et al.
Tetrahedron Letters, 45(16), 3253-3256 (2004)
Molecular-radical catalysis of the chain reaction of quinone imine with hydroquinone under the action of aromatic amines.
Varlamov VT.
Kinetics and Catalysis, 45(5), 662-675 (2004)
Dimethyldiphenylamino-substituted carbazoles as electronically active molecular materials.
Tomkeviciene A, et al.
Dyes and Pigments, 96(2), 574-580 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service