Skip to Content
Merck
All Photos(1)

Documents

762040

Sigma-Aldrich

Dibenzocyclooctyne-sulfo-N-hydroxysuccinimidyl ester

for Copper-free Click Chemistry

Synonym(s):

DBCO-sulfo-NHS ester, DBCO-sulfo-SE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N2NaO8S
Molecular Weight:
532.50
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: click chemistry
reagent type: linker

mp

240-260 °C

storage temp.

−20°C

SMILES string

O=C(CCCCC(ON(C(CC1S(=O)(O[Na])=O)=O)C1=O)=O)N2CC3=C(C=CC=C3)C#CC4=C2C=CC=C4

InChI

1S/C25H22N2O8S.Na/c28-22(11-5-6-12-24(30)35-27-23(29)15-21(25(27)31)36(32,33)34)26-16-19-9-2-1-7-17(19)13-14-18-8-3-4-10-20(18)26;/h1-4,7-10,21H,5-6,11-12,15-16H2,(H,32,33,34);/q;+1/p-1

InChI key

QXKHPYSXPCJSPR-UHFFFAOYSA-M

Application

Water soluble succinimidyl ester/NHS functionalized cyclooctyne derivative for incorporation of the cyclooctyne moiety into amine containing compounds or biomolecules. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage.
Dibenzocyclooctyne-sulfo-N-hydroxysuccinimidyl ester may be used for the covalent modification of nano-sized metal-organic frameworks (NMOFs) with nucleic acids via click chemistry during the synthesis stimuli-responsive DNA-gated NMOFs. These NMOFs can act as carriers of a fluorescent agent or anticancer drug, making them highly useful in targeted drug release system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATP?Responsive Aptamer?Based Metal?Organic Framework Nanoparticles (NMOFs) for the Controlled Release of Loads and Drugs.
Chen WH, et al.
Advances in Functional Materials (2017)
Switchable supramolecular catalysis using DNA-templated scaffolds.
Garcia MAA, et al.
Chemical Communications (Cambridge, England), 52(10), 2153-2156 (2016)
Optically Transparent Dense Colloidal Gels.
Zupkauskas M, et al.
Chemical Science (2017)
Instructing cells with programmable peptide DNA hybrids.
Freeman R, et al.
Nature Communications, 8 (2017)
Chen Chen et al.
Small (Weinheim an der Bergstrasse, Germany), 15(43), e1901014-e1901014 (2019-09-04)
Exosomes are endosome-derived vesicles enriched in body fluids such as urine, blood, and saliva. So far, they have been recognized as potential biomarkers for cancer diagnostics. However, the present single-variate analysis of exosomes has greatly limited the accuracy and specificity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service