Skip to Content
Merck
All Photos(1)

Key Documents

76153

Sigma-Aldrich

PEGA resin

extent of labeling: ~0.4 mmol/g loading

Synonym(s):

Poly[acryloyl-bis(aminopropyl)polyethylene glycol]

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.22

product line

PEGA

Quality Level

form

crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.4 mmol/g loading

impurities

90% methanol

particle size

150-300 μm

functional group

amine

Looking for similar products? Visit Product Comparison Guide

General description

PEGA resins are a hydrophilic acrylamide-PEG co-polymer providing a really extensive and more uniform swelling in a wide range of solvents (from water, methanol, ethanol to tetrahydrofuran, acetonitrile and toluene). The extreme swelling volume of this support in the standard peptide synthesis solvents DCM and DMF together with their good stability make them ideally suited for the batch as well as for continuous-flow syntheses.

Application

Polyacrylamide resin possessing long polyethylene glycol spacers giving extremely high swell resin may be used for:
  • immobilizing enzymes in aqueous solution
  • affinity purification of small-molecule binding proteins
  • synthesis of peptide libraries with fluorescently labeled proteins

Packaging

Sold on basis of weight of dry resin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Large scale modification of biomolecules using immobilized sortase A from Staphylococcus aureus
Steinhagen M, et al.
Bioorganic & Medicinal Chemistry, 21(12), 3504-3510 (2013)
A PEGA resin for use in the solid-phase chemical?enzymatic synthesis of glycopeptides.
Meldal M, et al.
Journal of the Chemical Society. Chemical Communications, 16, 1849-1850 (1994)
Synthesis, characterization and biocompatibility of PEGA resins.
Auzanneau FI, et al.
Journal of Peptide Science, 1(1), 31-44 (1995)
PEGA supports for combinatorial peptide synthesis and solid-phase enzymatic library assays.
Renil M, et al.
Journal of Peptide Science, 4(3), 195-195 (1998)
Catch and release of concanavalin A by a mannose-immobilized photoaffinity PEGA resin coupled with a cleavable disulfide linker.
Kuramochi K, et al.
Bioscience, Biotechnology, and Biochemistry, 79(12), 1946-1953 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service