Skip to Content
Merck
All Photos(1)

Documents

760811

Sigma-Aldrich

Bis[2-(trimethylsilyl)ethyl] N,N-diisopropylphosphoramidite

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H40NO2PSi2
CAS Number:
Molecular Weight:
365.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.4517

bp

286.2-296.6 °C

density

0.8887 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)N(C(C)C)P(OCC[Si](C)(C)C)OCC[Si](C)(C)C

InChI

1S/C16H40NO2PSi2/c1-15(2)17(16(3)4)20(18-11-13-21(5,6)7)19-12-14-22(8,9)10/h15-16H,11-14H2,1-10H3

InChI key

UCVPTNNSVPPRRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphitylation agent used in the preparation of oligonucleotides

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service