Skip to Content
Merck
All Photos(2)

Documents

756423

Sigma-Aldrich

Strontium nitride

powder, -60 mesh, 99.5%, Ba: <1%

Synonym(s):

Tristrontium dinitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr3N2
CAS Number:
Molecular Weight:
290.87
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5%

form

powder

particle size

-60 mesh

SMILES string

[Sr]=N[Sr]N=[Sr]

InChI

1S/2N.3Sr

InChI key

PVVTZMTUGZOOIR-UHFFFAOYSA-N

Application

Strontium nitride (Sr3N2) can be used in the synthesis of ternary nitride oxide with molybdenum nitride under a low partial pressure of oxygen. It can also be used in the synthesis of barium-strontium silicate-based hollow structures for potential usage in drug delivery, catalysis, and sensors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruan, J.; et al.
Journal of the American Ceramic Society. American Ceramic Society, 94, 536-536 (2011)
Synthesis and crystal structure of a new strontium nitridomolybdate oxide, Sr 4 [MoN 4] O
O?Meara PM, et al.
Journal of the Chemical Society, Part 1, 633-638 (2000)
Sharma, S.B.; Paliwal, P.; Kumar, M.
Journal of Physics and Chemistry of Solids, 51, 35-35 (1990)
Gregory, D.H.; et al.
J. Alloy Compounds, 317, 237-237 (2001)
Template-engaged solid-state synthesis of barium-strontium silicate hexagonal tubes
Chen X and Kim W
Journal of alloys and compounds, 647(5), 1128-1135 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service