753947
ICMA
97% (HPLC)
Synonym(s):
1′,4′-Dihydro-naphtho[2′,3′:1,2][5,6]fullerene-C60, C60 derivative, indene-C60 monoadduct
About This Item
Recommended Products
Quality Level
Assay
97% (HPLC)
form
solid
loss
0.5 wt. %, 390 °C
mp
320-340 °C
Orbital energy
LUMO 3.84 eV (CV)
OPV Device Performance
ITO/PEDOT:PSS/P3HT: ICMA (1:1)/Al
ITO/PEDOT:PSS/P3HT:ICMA(1:1)/LiF/Al
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service