Skip to Content
Merck
All Photos(4)

Key Documents

745405

Sigma-Aldrich

Sodium 1,1-difluoroethanesulfinate

Synonym(s):

Baran DFES Reagent, Baran Difluoroethylation Reagent, DFES-Na Reagent, Sodium 1,1-difluoroethylsulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3F2NaO2S
CAS Number:
Molecular Weight:
152.10
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: C-C Bond Formation
reaction type: Fluorinations
reagent type: catalyst
reaction type: C-H Activation

reagent type: diversification reagent

mp

>300 °C

functional group

fluoro
sulfinic acid

SMILES string

[Na+].CC(F)(F)S([O-])=O

InChI

1S/C2H4F2O2S.Na/c1-2(3,4)7(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

YAUVUSSCQUHHFC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

DFES-Na provides a direct approach to fluorinated heteroarylether bioisosteres by appending a 1,1-difluoroethyl group operationally simple and robust protocol. This reagent was developed by the Baran Group and is one of several reagents for the direct alkylation of heterocycles.

Learn More at the Professor and Product Portal of Professor Phil S. Baran.
Baran Diversinates(TM) Lead Diversification and Metabolism Prediction Flyer

Linkage

Frequently Asked Questions are available for this Product.

Other Notes

Previously sold under product number L511404

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct synthesis of fluorinated heteroarylether bioisosteres.
Qianghui Zhou et al.
Angewandte Chemie (International ed. in English), 52(14), 3949-3952 (2013-03-06)
Fionn O'Hara et al.
Journal of the American Chemical Society, 135(32), 12122-12134 (2013-07-19)
Radical addition processes can be ideally suited for the direct functionalization of heteroaromatic bases, yet these processes are only sparsely used due to the perception of poor or unreliable control of regiochemistry. A systematic investigation of factors affecting the regiochemistry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service