Skip to Content
Merck
All Photos(1)

Key Documents

739928

Sigma-Aldrich

3-epi-25-Hydroxyvitamin D3 solution

50 μg/mL in ethanol, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Assay

98% (CP)

form

solution

concentration

50 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

mp

-144.0 °C

application(s)

detection

storage temp.

−20°C

SMILES string

C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@H](O)CCC3=C

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-UEGFJKGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-epi-25-Hydroxyvitamin D3 (C3-epimer) is a metabolite of vitamin D3. It is an epimer of 25-hydroxyvitamin D3 which differs only at C-3 position wherein -OH group at C-3 carbon is in the α orientation rather than in the β orientation.[1]

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overestimation of 25-hydroxyvitamin D3 by increased ionisation efficiency of 3-epi-25-hydroxyvitamin D3 in LC-MS/MS methods not separating both metabolites as determined by an LC-MS/MS method for separate quantification of 25-hydroxyvitamin D3, 3-epi-25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 in human serum
van den Ouweland JMW, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 967, 195-202 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service