Skip to Content
Merck
All Photos(1)

Key Documents

737860

Sigma-Aldrich

Sodium molybdate

anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis

Synonym(s):

Disodium molybdate, Natrium molybdat, Sodium Molybdenum Oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2MoO4
CAS Number:
Molecular Weight:
205.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.9% trace metals basis

form

powder

impurities

≤2000.0 ppm Trace Metal Analysis

particle size

−100 mesh

mp

687 °C (lit.)

density

3.78 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Na+].[Na+].[O-][Mo]([O-])(=O)=O

InChI

1S/Mo.2Na.4O/q;2*+1;;;2*-1

InChI key

TVXXNOYZHKPKGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium molybdate is a white powder that is an environmentally friendly non-oxidizing corrosion inhibitor and Mo source which is widely used in rechargeable batteries and supercapacitors.

Application

Sodium molybdate can be used as:
  • A precursor to prepare MoS2-based electrode materials for sodium-ion batteries with high initial Coulombic efficiency (ICE), high-rate performance, and long cycle life.
  • An electrolyte additive in energy storage devices such as supercapacitors, to prevent the corrosion of stainless-steel collectors and enhance the performance.
  • A source of molybdenum to synthesize MoS2 quantum dots that can be used in fluorescence sensors for ascorbic acid detection.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium molybdate-an additive of choice for enhancing the performance of AC/AC electrochemical capacitors in a salt aqueous electrolyte
Q Abbas, et al.
Faraday Discussions, 172, 199-214 (2014)
A simple and high-performance supercapacitor based on nitrogen-doped porous carbon in redox-mediated sodium molybdate electrolyte
Kanjun Sun, et al
Electrochimica Acta, 158, 361-367 (2015)
Switch-on fluorescence sensor for ascorbic acid detection based on MoS2 quantum dots-MnO2 nanosheets system and its application in fruit samples
Ya-Li Xu, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 28, 338-344 (2017)
Kimberly M Stroka et al.
Oncotarget, 8(33), 54004-54020 (2017-09-15)
Obscurins are a family of RhoGEF-containing proteins with tumor and metastasis suppressing roles in breast epithelium. Downregulation of giant obscurins in normal breast epithelial cells leads to reduced levels of active RhoA and of its downstream effectors. Herein, we elucidate
MoS2 nanosheets vertically aligned on carbon paper: a freestanding electrode for highly reversible sodium-ion batteries
Xiuqiang Xie, et al.
Advanced Energy Materials, 6, 1502161-1502161 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service