Skip to Content
Merck
All Photos(1)

Key Documents

736287

Sigma-Aldrich

PEDOT

bis-PEG, lauryl terminated, 0.8 wt. % (propylene carbonate dispersion), contains perchlorate as dopant

Synonym(s):

Aedotron C3-PC, C12-PEG-block-PEDOT-block-PEG-C12, PEDOT:PEG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

product name

Poly(3,4-ethylenedioxythiophene), bis-poly(ethyleneglycol), lauryl terminated, 0.8 wt. % (dispersion in propylene carbonate), contains perchlorate as dopant

form

liquid (dispersion)

Quality Level

contains

perchlorate as dopant

composition

acetonitrile, 4-8 wt. %
Aedotron TM-C3 polymer, 0.2-0.9 wt. %
propietary processing additive, 0.1-0.7 wt. %
propylene carbonate, 90-95 wt. %
propylene glycol, 0.0-0.3 wt. %

concentration

0.6-1.1 wt. %
0.8 wt. % (dispersion in propylene carbonate)

refractive index

n20/D 1.411

particle size

200-600 nm (in suspension)

conductivity

10-45 S/cm (bulk)

density

1.141 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Aedotron polymers are solvent-dispersable forms of PEDOT. These are copolymers of doped PEDOT and a flexible, soluble polymer such as poly(ethylene glycol) (PEG). These colloidal dispersions are stabilized by the highly solvated PEG chains which sterically limit the aggregation of the PEDOT blocks. Aedotron polymers are neither acidic nor corrosive, it easily disperses in polar aprotic solvents.
High transparency and high conductivity dispersion in a non-volatile solvent

Application

A single-pass line has the following characteristics:
  • Printed line width, 120 μm
  • Printed line cross-sectional area, 14.1μm2
  • Printed line average height, 70.6 ηm
  • Sheet resistance, 2.0 kOhms/Square
  • Resistivity, 14,021 μOhms·cm (71 S/cm)
  • Fairly good optical clarity
Conducting polymer dispersion suitable for making conducting films, anti-static layers and transparent electrodes for electroluminescent lamps. 1,2 Dichlorobenzene can be used to cast robust films on many polymeric substrates, but should be tested in each case.

Features and Benefits

The block copolymer structure renders the conducting polymer PEDOT highly dispersible in organic solvents. dichlorobenzene is compatible with many coatings and polymers formulations.

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling and aggregation will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Other Notes

Not for spin-cast

Legal Information

Aedotron is a trademark of TDA Research, Inc.
Aedotron is a trademark of TDA Research, Inc.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.0 °F

Flash Point(C)

37.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service