Skip to Content
Merck
All Photos(1)

Key Documents

735973

Sigma-Aldrich

N,N′-Dimethyl N,N′-di(4-pyridinyl)thiuram disulfide

Synonym(s):

Bis[methyl-(4-pyridinyl)thiocarbamoyl] disulfide, Bis[methyl-(4-pyridinyl)thiocarbamyl] disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4S4
CAS Number:
Molecular Weight:
366.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

mp

118-128 °C

SMILES string

CN(C(=S)SSC(=S)N(C)c1ccncc1)c2ccncc2

InChI

1S/C14H14N4S4/c1-17(11-3-7-15-8-4-11)13(19)21-22-14(20)18(2)12-5-9-16-10-6-12/h3-10H,1-2H3

InChI key

PVZGPMRTEJHLMS-UHFFFAOYSA-N

General description

Precursor for the synthesis of novel switchable RAFT agents for controlled radical polymerization. Chain Transfer Agent (CTA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service