Skip to Content
Merck
All Photos(1)

Key Documents

735353

Sigma-Aldrich

Lithium 2,2,6,6-tetramethylpiperidide

95%

Synonym(s):

2,2,6,6-Tetramethylpiperidine lithium salt (1:1), LTMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18LiN
CAS Number:
Molecular Weight:
147.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

SMILES string

[Li]N1C(C)(C)CCCC1(C)C

InChI

1S/C9H18N.Li/c1-8(2)6-5-7-9(3,4)10-8;/h5-7H2,1-4H3;/q-1;+1

InChI key

ANYSGBYRTLOUPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium 2,2,6,6-tetramethylpiperidide is a strong base and can be used:
  • For the synthesis of enamines from terminal epoxides through trans-α-lithiated epoxide as an intermediate.
  • For ortholithiation of arenes such as 1,3-bis(trifluoromethyl)benzene, 4,4-dimethyl-2-phenyl-2-oxazoline, 1,4-bis(trifluoromethyl)benzene and 1,3-dimethoxybenzene.
  • In combination with Lewis donor ligand, N,N,N′,N′-tetramethylethylenediamine (TMEDA) for deprotonative metalation of methoxy-substituted arenes.{21]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkenes from terminal epoxides using lithium 2,2,6,6-tetramethylpiperidide and organolithiums or grignard reagents.
Hodgson DM, et al.
Journal of the American Chemical Society, 126(39), 12250-12251 (2004)
Deprotonative metalation of methoxy-substituted arenes using lithium 2, 2, 6, 6-tetramethylpiperidide: Experimental and computational study.
Akimoto G, et al.
The Journal of Organic Chemistry, 83(21), 13498-13506 (2018)
Case for Lithium Tetramethylpiperidide-Mediated Ortholithiations: Reactivity and Mechanisms.
Mack KA and Collum DB
Journal of the American Chemical Society, 140(14), 4877-4883 (2018)
Martin-Louis Y Riu et al.
Science advances, 6(13), eaaz3168-eaaz3168 (2020-04-02)
This exploratory synthesis investigation was undertaken to determine the viability of replacing a single carbon vertex with another p-block element in a highly strained tetrahedrane molecule. Phosphorus was selected for this purpose because the stable molecular form of elemental phosphorus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service