Skip to Content
Merck
All Photos(1)

Key Documents

730343

Sigma-Aldrich

Pyrazine-2-thiocarboxamide

97%

Synonym(s):

Pyrazine-2-carbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3S
CAS Number:
Molecular Weight:
139.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

195-200 °C

functional group

amine

SMILES string

NC(=S)c1cnccn1

InChI

1S/C5H5N3S/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)

InChI key

LIURPUMROGYCLW-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service