Skip to Content
Merck
All Photos(1)

Documents

72568

Sigma-Aldrich

Nitrilotri(methylphosphonic acid)

≥95% (T)

Synonym(s):

ATMP, Aminotris(methylenephosphonic acid), [Bis(phosphonomethyl)amino]methylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N[CH2PO(OH)2]3
CAS Number:
Molecular Weight:
299.05
Beilstein:
1715724
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (T)

form

crystals

mp

~215 °C (dec.)

SMILES string

OP(O)(=O)CN(CP(O)(O)=O)CP(O)(O)=O

InChI

1S/C3H12NO9P3/c5-14(6,7)1-4(2-15(8,9)10)3-16(11,12)13/h1-3H2,(H2,5,6,7)(H2,8,9,10)(H2,11,12,13)

InChI key

YDONNITUKPKTIG-UHFFFAOYSA-N

Application

Nitrilotri(methylphosphonic acid) is a common chelating agent used in synthetic chemistry. Some of the other applications include:
  • Preparation of hexagonal porous three-dimensional structures encapsulating a template, layered structures with intercalated templates or linear polymers.
  • Synthesis of metal-organic frameworks in combination with uranyl nitrate.
  • Preparation of ingredient of anticorrosive protective coatings on the steel surface.
  • ATMP can also be employed as a scale inhibitor during squeeze treatments in oilfield operations.

Other Notes

Powerful complexing agent

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and properties of nitrilo-tris (methylenephosphonato)-triaquairon (II){Fe [μ-NH (CH 2 PO 3 H) 3](H 2 O) 3}, as an ingredient of anticorrosive protective coatings on the steel surface.
Somov N V, et al.
Crystallography Reports, 60(6), 853-859 (2015)
Nitrilotris (methylenephosphonato) potassium K [μ6-NH (CH2PO3) 3H4]: Synthesis, structure, and the nature of the K?O chemical bond.
Somov N V, et al.
Crystallography Reports, 61(4), 606-610 (2016)
Complexes formed between nitrilotris (methylenephosphonic acid) and M2+ transition metals: isostructural organic? inorganic hybrids.
Cabeza A, et al.
Inorganic Chemistry, 41(9), 2325-2333 (2002)
[Hygienic establishment of maximum permissible concentrations of nitrilotrimethylphosphonic acid and a copper-containing complex in reservoir water].
R A Suleĭmanov et al.
Gigiena i sanitariia, (2)(2), 67-68 (1984-02-01)
Yuta Suzuki et al.
Nucleic acids symposium series (2004), (52)(52), 379-380 (2008-09-09)
Only one phosphodiester linkage in long DNA was hydrolyzed simply by using two components: oligonucleotide-multiphosphonate conjugates ("gapmers") and Ce(IV)/EDTA (Fig. 1).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service