Skip to Content
Merck
All Photos(1)

Key Documents

724831

Sigma-Aldrich

Poly(N-isopropylacrylamide), amine terminated

average Mn 5,500 (T)

Synonym(s):

NIPAM polymer, PNIPAM-NH2, Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(C6H11NO)nSCH2CH2NH2
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 5,500 (HCl titration)

Looking for similar products? Visit Product Comparison Guide

Application

Novel polymer for development of thermosensitive coated micro/nano materials including thermoresponsive polymeric drug delivery systems. The amine functional group can be used to conjugate a variety of biomolecules to the polymer chain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service