Skip to Content
Merck
All Photos(2)

Key Documents

721034

Sigma-Aldrich

5-Chloro-3-pyridineboronic acid

≥95%

Synonym(s):

(3-Chloro-5-pyridyl)boronic acid, (3-Chloropyridin-5-yl)boronic acid, (5-Chloropyridin-3-yl)boronic acid, 3-Chloropyridine-5-boronic acid, 5-Chloropyridine-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BClNO2
CAS Number:
Molecular Weight:
157.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

255-260 °C

storage temp.

2-8°C

SMILES string

OB(O)c1cncc(Cl)c1

InChI

1S/C5H5BClNO2/c7-5-1-4(6(9)10)2-8-3-5/h1-3,9-10H

InChI key

NJXYBTMCTZAUEE-UHFFFAOYSA-N

Application

5-Chloro-3-pyridineboronic acid can be used as an intermediate:
  • In the preparation of positron emission tomography (PET) radioligand [11C]MK-1064, which is applicable in the orexin-2 receptor imaging.
  • In the synthesis of orexin 2 receptor antagonist (2-SORA) MK-1064.
  • In the preparation of pyridyl-cycloalkyl derived microsomal prostaglandin E synthase-1 inhibitors.
  • In the synthesis of ATAD2 bromodomain inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and optimization of pyridyl-cycloalkyl-carboxylic acids as inhibitors of microsomal prostaglandin E synthase-1 for the treatment of endometriosis
Koppitz M, et al.
Bioorganic & Medicinal Chemistry Letters, 29(18), 2700-2705 (2019)
Discovery of 5″-Chloro-N-[(5,6-dimethoxypyridin-2-yl) methyl]-2, 2′: 5′, 3″-terpyridine-3′-carboxamide (MK-1064): A Selective Orexin 2 Receptor Antagonist (2-SORA) for the Treatment of Insomnia
Roecker AJ, et al.
ChemMedChem, 9(2), 311-322 (2014)
Synthesis of [11C] MK-1064 as a new PET radioligand for imaging of orexin-2 receptor
Gao M, et al.
Bioorganic & Medicinal Chemistry Letters, 26(15), 3694-3699 (2016)
Fragment-based discovery of low-micromolar ATAD2 bromodomain inhibitors
Demont EH, et al.
Journal of Medicinal Chemistry, 58(14), 5649-5673 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service