Skip to Content
Merck
All Photos(1)

Documents

720186

Sigma-Aldrich

Bisphenol-A-(diphenyl-13C12)

99 atom % 13C, 98% (CP)

Synonym(s):

2,2-Bis(4-hydroxyphenyl-13C6)propane, 4,4′-Isopropylidenediphenol-13C12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(13C6H4OH)2
CAS Number:
Molecular Weight:
240.20
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

crystals

technique(s)

mass spectrometry (MS): suitable

mp

158-159 °C (lit.)

mass shift

M+12

SMILES string

CC(C)([13c]1[13cH][13cH][13c](O)[13cH][13cH]1)[13c]2[13cH][13cH][13c](O)[13cH][13cH]2

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1

InChI key

IISBACLAFKSPIT-ZGFLEIAISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service