71716
Sodium D-lactate
≥99.0% (NT)
Synonym(s):
(R)-2-Hydroxypropionic acid sodium salt, D-Lactic acid sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06
Beilstein:
5454721
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99.0% (NT)
form
powder
optical activity
[α]20/D +12.5±0.5°, c = 1% in H2O
optical purity
enantiomeric ratio: ≥99:1 (enzymatic)
functional group
hydroxyl
SMILES string
[Na+].C[C@@H](O)C([O-])=O
InChI
1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m1./s1
InChI key
NGSFWBMYFKHRBD-HSHFZTNMSA-M
General description
Sodium D-lactate is the sodium salt of D-lactic acid. It is formed as an intermediate during the carbohydrate metabolism in muscle and liver.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Metabolism of Sodium d-Lactate: I. Utilization of Intravenously Injected Sodium d-Lactate by Normal Persons.
Soffer L J, et al.
Archives of Internal Medicine, 60(5), 876-881 (1937)
Joel Frandsen et al.
Scientific reports, 7(1), 5126-5126 (2017-07-13)
Oxidative stress is damaging to cells and contributes to aging and neurodegenerative disease. This state is mediated by production of imbalanced molecules, and reactive dicarbonyl compounds - mainly methylglyoxal. The glyoxalase pathway is an antioxidant defense system utilized to detoxify
Xiaoxia Zhang et al.
Journal of industrial microbiology & biotechnology, 46(7), 899-909 (2019-04-10)
Lactoyl-CoA is critical for the biosynthesis of biodegradable and biocompatible lactate-based copolymers, which have wide applications. However, reports on acetyl-CoA: lactate CoA-transferases (ALCTs) are rare. To exploit novel ALCTs, amino acid sequence similarity searches based on the CoA-transferases from Clostridium
Braedon McDonald et al.
Cell host & microbe, 28(5), 660-668 (2020-08-19)
Eradication of pathogens from the bloodstream is critical to prevent disseminated infections and sepsis. Kupffer cells in the liver form an intravascular firewall that captures and clears pathogens from the blood. Here, we show that the catching and killing of
Yusuke Toyoda et al.
Nature communications, 8(1), 1266-1266 (2017-11-04)
To divide, most animal cells drastically change shape and round up against extracellular confinement. Mitotic cells facilitate this process by generating intracellular pressure, which the contractile actomyosin cortex directs into shape. Here, we introduce a genome-scale microcantilever- and RNAi-based approach
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service