Skip to Content
Merck
All Photos(1)

Key Documents

715581

Sigma-Aldrich

Pyrrolidine-3-carboxylic acid hydrochloride

95%

Synonym(s):

β-Proline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10ClNO2
CAS Number:
Molecular Weight:
151.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
95%

Quality Level

Assay

95%

form

solid

mp

33-38 °C

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

Cl.OC(=O)C1CCNC1

InChI

1S/C5H9NO2.ClH/c7-5(8)4-1-2-6-3-4;/h4,6H,1-3H2,(H,7,8);1H

InChI key

OYCLYMMIZJWYJG-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service