Skip to Content
Merck
All Photos(1)

Documents

710660

Sigma-Aldrich

(11bS)-4,5-Dihydro-3H-dinaphtho[2,1-c:1′,2′-e]phosphepine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17P
Molecular Weight:
312.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

optical activity

[α]22/D +368°, c = 0.5 in chloroform

mp

158-162 °C

SMILES string

C1PCc2ccc3ccccc3c2-c4c1ccc5ccccc45

InChI

1S/C22H17P/c1-3-7-19-15(5-1)9-11-17-13-23-14-18-12-10-16-6-2-4-8-20(16)22(18)21(17)19/h1-12,23H,13-14H2

InChI key

ZOEOSXZNQTXHDI-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service