Skip to Content
Merck
All Photos(1)

Key Documents

704288

Sigma-Aldrich

Lanthanum strontium cobalt ferrite

greener alternative

LSCF 6428

Synonym(s):

LCSF, Lanthanum strontium cobalt iron oxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

powder

composition

La0.6Sr0.4Co0.2Fe0.8O3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

4-8 m2/g

particle size

0.7-1.1 μm

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Lanthanum strontium cobalt ferrite (LSCF 6428) is a promising cathode material, which has a low operating temperature. It has a high ionic and electronic conductivity with a high electrocatalytic activity.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

LSCF- 6428 is a mixed ionic and electron conducting pervoskite. The ionic conductivity is lower than YSZ although maintaining high conductivity. X-ray diffraction and electrochemical measurements of an operational LSCF-6428 cathode was conducted to study its lattice expansion during use. LSCF-6428 can be prepared by solid state reaction of individual cation oxides.
LSCF-6428 may be used as the cathode in solid oxide fuel cells (SOFCs).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lattice expansion of LSCF-6428 cathodes measured by in situ XRD during SOFC operation.
Hardy JS, et al.
Journal of Power Sources, 198, 76-82 (2012)
Electrochemical performance and stability of lanthanum strontium cobalt ferrite oxygen electrode with gadolinia doped ceria barrier layer for reversible solid oxide fuel cell
Fan H, et al.
Journal of Power Sources, 268, 634-639 (2014)
Preparation of an asymmetric perovskite-type membrane and its oxygen permeability
Jin W, et al.
Journal of Membrane Science, 185(2), 237-243 (2001)
Advances in Materials for Solid Oxide Fuel Cells
Gorte RJ, et al.
Material Matters, 5(4) null
Ultra-thin nanocrystalline lanthanum strontium cobalt ferrite (La0. 6Sr0. 4Co0. 8Fe0. 2O3- delta) films synthesis by RF-sputtering and temperature-dependent conductivity studies
Lai B, et al.
Journal of Power Sources, 186(1), 115-122 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service