Skip to Content
Merck
All Photos(4)

Key Documents

704121

Sigma-Aldrich

Carbon nanotube, single-walled

(7,6) chirality, ≥90% carbon basis (≥77% as carbon nanotubes), 0.83 nm average diameter, avg. no. of layers, 1

Synonym(s):

CHASM, CNT, Signis® SG76, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

description

G/D Ratio: ≥15 (Raman 633nm)
Median length: 1 μm

Assay

≥90% carbon basis (≥77% as carbon nanotubes)

form

powder (freeze-dried)

feature

avg. no. of layers 1

manufacturer/tradename

Signis® SG76

surface area

≥700 m2/g

impurities

≤5 wt. % Moisture content

average diameter

0.83 nm

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

bulk density

0.1 g/cm3

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced using CHASM′s patented CoMoCAT technology, Signis® SG76 is selectively enriched in (7,6) chirality.
Tube diameter determined from an optical absorbance spectrum and AFM.

Application

Carbon nanotube, single-walled (SWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, thin film conductors, field effect transistors (FETs), biosensor, gas sensor, supercapacitor and nanomechanical resonators.
Suitable for use in coatings for printed electronics, photovoltaics, medical applications.SWNTs (7,6 chirality) was used to prepare SWNT buckypapers, which were used as electrocatalyst supports for the electro-oxidation of methanol.SWNTs on poly methyl methacrylate (PMMA) substrates may be used as transducers for electrochemical microfluidic sensing.

Preparation Note

CoMoCAT Catalytic Chemical Vapor Deposition (CVD) Method

Legal Information

CHASM is a trademark of Chasm Advanced Materials
CoMoCAT is a trademark of Chasm Advanced Materials
Signis is a registered trademark of Chasm Advanced Materials

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral-Selective Growth of Single-Walled Carbon Nanotubes on Lattice-Mismatched Epitaxial Cobalt Nanoparticles.
He M, et al.
Scientific Reports, 3(1460) null
Two-dimensional gel electrophoresis and mass spectrometry in studies of nanoparticle-protein interactions.
Karlsson H, et al.
Gel Electrophoresis-Advanced Techniques (2012)
Natalia Vladimirovna Kamanina et al.
Sensors (Basel, Switzerland), 18(9) (2018-09-13)
A potassium bromide (KBr) material, which has been widely used as the key element in Fourier spectrometers and as the output window of the IR-lasers, was studied via applying carbon nanotubes in order to modify the potassium bromide surface. The
etal impurities provide useful tracers for identifying exposures to airborne single-wall carbon nanotubes released from work-related processes
Rasmussen PE, et al.
Journal of Physics. Conference Series, 429 (2013)
Electrochemical behaviour of different redox probes on single wall
carbon nanotube buckypaper-modified electrodes
Sieben JM, et al.
Electrochimica Acta, 135, 404-411c-404-411c (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service