Skip to Content
Merck
All Photos(1)

Documents

704067

Sigma-Aldrich

ε-Caprolactone

97%

Synonym(s):

2-Oxepanone, 6-Caprolactone monomer, 6-Hexanolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
106919
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.9 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

97-98 °C/15 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCO1

InChI

1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2

InChI key

PAPBSGBWRJIAAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ε-caprolactone is a cyclic ester. Ring opening polymerization of ε-caprolactone initiated by, tributylin n butoxide, ethyl magnesium bromide, or samarium acetate or heteropolyacid, leads to the formation of poly caprolactone (PCL).

Application

ε-caprolactone is popularly used in the preparation of poly caprolactone (PCL).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanzhu Guo et al.
Carbohydrate polymers, 92(1), 77-83 (2012-12-12)
Self-associating cellulose-graft-poly(ɛ-caprolactone) (cellulose-g-PCL) copolymers were successfully synthesized via homogeneous ring-opening polymerization (ROP) of ɛ-CL onto softwood dissolved pulp substrate in ionic liquid 1-N-butyl-3-methylimidazolium chloride ([Bmim]Cl). An organic catalyst N,N-dimethylamino-4-pyridine (DMAP) was compared with the traditional metal-based catalyst (Sn(Oct)(2)) as the
Leonardus Kresna Widjaja et al.
Journal of the mechanical behavior of biomedical materials, 6, 80-88 (2012-02-04)
A series of triblock copolymers comprising end block of PLLA modified with PCL, and random copolymer of PCL and PTMC as soft segment were synthesized. DSC data show that PCL disrupted the crystallinity of PLLA, making the hard block to
Sepideh Jalilzadeh-Tabrizi et al.
Biopreservation and biobanking, 16(2), 66-76 (2018-01-23)
Emu oil (EO) with anti-inflammatory, antioxidative, and wound healing properties can be blended for preparing bioactive nanofibrous scaffold. Adipose tissue-derived stem cells (ADSCs) are promising candidates for tissue engineering, and preserving their stemness potential is vital for further therapeutic applications.
Eliza Li Shan Fong et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(16), 6500-6505 (2013-04-12)
The pronounced biological influence of the tumor microenvironment on cancer progression and metastasis has gained increased recognition over the past decade, yet most preclinical antineoplastic drug testing is still reliant on conventional 2D cell culture systems. Although monolayer cultures recapitulate
Ring-opening polymerization of e-caprolactone initiated by heteropolyacid
Cheng G, et al.
Journal of Polymer Research, 17(6), 847-851 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service