Skip to Content
Merck
All Photos(1)

Key Documents

694371

Sigma-Aldrich

2-Fluoropropionic acid

97%

Synonym(s):

α-Fluoropropionic acid, (±)-2-Fluoropropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5FO2
CAS Number:
Molecular Weight:
92.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.383

bp

66-67 °C/30 mmHg

density

1.181 g/mL at 25 °C

functional group

carboxylic acid
fluoro

SMILES string

CC(F)C(O)=O

InChI

1S/C3H5FO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)

InChI key

ZVZPFTCEXIGSHM-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

180.0 °F

Flash Point(C)

82.22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junling Li et al.
Nuclear medicine and biology, 42(3), 283-291 (2014-12-30)
Bis(Zn-dipicolylamine (Zn-DPA)) coordination complexes represent a new class of synthetic small molecules that can target anionic phosphatidylserine (PS) in the apoptotic cells with high affinity and specificity. In this study, we labeled Zn-DPA and Cy7-Zn-DPA with different (18)F-prosthetic groups and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service