Skip to Content
Merck
All Photos(1)

Documents

693243

Sigma-Aldrich

(S)-Ru(OAc)2(SEGPHOS®)

Synonym(s):

Diacetato[(S)-(−)-5,5′-bis(diphenylphosphino)-4,4′-bi-1,3-benzodioxole]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H34O8P2Ru
CAS Number:
Molecular Weight:
829.73
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

storage temp.

2-8°C

InChI

1S/C38H28O4P2.2C2H4O2.Ru/c1-5-13-27(14-6-1)43(28-15-7-2-8-16-28)33-23-21-31-37(41-25-39-31)35(33)36-34(24-22-32-38(36)42-26-40-32)44(29-17-9-3-10-18-29)30-19-11-4-12-20-30;2*1-2(3)4;/h1-24H,25-26H2;2*1H3,(H,3,4);/q;;;+2/p-2

InChI key

BHGLLIGZFQVMBJ-UHFFFAOYSA-L

Application

(S)-Ru(OAc)2(SEGPHOS®) can be used as a catalyst:
  • To prepare highly chemo, enantio, and diastereoselective primary β-amino lactams by asymmetric reductive amination of racemic β-keto lactams.
  • To synthesize chiral primary diarylmethylamines and sterically bulky benzylamines from diaryl and sterically hindered ketones via asymmetric reductive amination reaction.
  • For the conversion of levulinic acid to optically active γ-valerolactone via asymmetric hydrogenation reaction.

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct asymmetric reduction of levulinic acid to gamma-valerolactone: synthesis of a chiral platform molecule
Tukacs JM, et al.
Green Chemistry, 17(12), 5189-5195 (2015)
Dynamic Kinetic Asymmetric Reductive Amination: Synthesis of Chiral Primary ?-Amino Lactams
Lou Y, et al.
Angewandte Chemie (International ed. in English), 57(43), 14193-14197 (2018)
Ruthenium-Catalyzed Direct Asymmetric Reductive Amination of Diaryl and Sterically Hindered Ketones with Ammonium Salts and H2
Hu L, et al.
Angewandte Chemie (International Edition in English), 132(13), 5359-5363 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service