Skip to Content
Merck
All Photos(2)

Key Documents

683167

Sigma-Aldrich

Chiralyst P461

Umicore

Synonym(s):

Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer, Di(μ-chloro)bis(norbornadiene)dirhodium(I), 2,5-Norbornadiene-rhodium(I) chloride dimer, Chloro(2,5-norbornadiene)rhodium(I) dimer, Rhodium(I) chloride 2,5-Norbornadiene complex dimer, [Rh(nbd)Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16Cl2Rh2
CAS Number:
Molecular Weight:
460.99
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

SMILES string

Cl[Rh].Cl[Rh].[CH]1[CH]C2[CH][CH]C1C2.[CH]3[CH]C4[CH][CH]C3C4

InChI

1S/2C7H8.2ClH.2Rh/c2*1-2-7-4-3-6(1)5-7;;;;/h2*1-4,6-7H,5H2;2*1H;;/q;;;;2*+1/p-2/t2*6-,7+;;;;

InChI key

RXDWVIOULPVOEO-SEGRDSFWSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for valence isomerization and N-heterocyclization; incombination with phosphorus ligands are catalysts for hydrogenationof many functional groups and hydroformylation; withchiral ligands are catalysts for asymmetric hydrogenation

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tani, K.; Tanigawa, E.; Tatsuno, Y.; Otsuka, S.
Journal of Organometallic Chemistry, 279, 87-87 (1985)
Becalski, A. G.; Cullen,W. R. et al.
Inorganic Chemistry, 30, 5002-5002 (1991)
Watanabe, Y.; Shim, S. C.; Mitsudo, T.
Bulletin of the Chemical Society of Japan, 54, 3460-3460 (1981)
Taylor, R. B.; Jennings, P. W.
Inorganic Chemistry, 20, 3997-3997 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service