Skip to Content
Merck
All Photos(1)

Documents

682624

Sigma-Aldrich

Nanoclay, surface modified

contains 35-45 wt. % dimethyl dialkyl (C14-C18) amine

Synonym(s):

Nanomer® I.44P, Nanomer® clay, Montmorillonite clay

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

contains

35-45 wt. % dimethyl dialkyl (C14-C18) amine

matrix

Montmorillonite clay base material

particle size

≤20 μm

Looking for similar products? Visit Product Comparison Guide

Application

Recommended for melt compounding with polypropylene (PP), polyethylene (PE), and ethylene vinyl acetate (EVA).
  • Improve modulus
  • Increase barrier
  • Enhance chemical resistance
  • Improve flame retardancy
  • Increase heat deflection temperature (HDT)

Legal Information

Nanomer® clays are products of Nanocor Corporation
Product of Nanocor®, Inc.
Nanomer is a registered trademark of Amcol International Corporation

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service