Skip to Content
Merck
All Photos(3)

Key Documents

682179

Sigma-Aldrich

2-(Bromomethyl)-6-methylpyridine

97%

Synonym(s):

6-Methyl-2-(bromomethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BrN
CAS Number:
Molecular Weight:
186.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

38-42 °C

SMILES string

Cc1cccc(CBr)n1

InChI

1S/C7H8BrN/c1-6-3-2-4-7(5-8)9-6/h2-4H,5H2,1H3

InChI key

WJFDCFHWFHCLIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Jung Wu et al.
Analytical and bioanalytical chemistry, 410(28), 7405-7414 (2018-09-08)
In the clinic, ethosuximide is commonly used to treat generalized absence seizures but has recently been repurposed for other diseases. Because of adverse effects and drug interactions, high-throughput therapeutic drug monitoring of ethosuximide is necessary. Microextraction is a simple, effective
Yi-Shan Li et al.
Talanta, 218, 121143-121143 (2020-08-17)
Dipeptidyl peptidase 4 (DPP-4) inhibitors are incretin-based medications used as oral antidiabetic agents for the treatment of type 2 diabetes. However, DPP-4 inhibitors produce side effects like acute pancreatitis, upper respiratory tract infection, nasopharyngitis, urinary tract infection, serious allergies, cardiovascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service