Skip to Content
Merck
All Photos(1)

Key Documents

679275

Sigma-Aldrich

Triethoxyvinylsilane

≥98%, deposition grade

Synonym(s):

(Triethoxysilyl)ethylene, Vinyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHSi(OC2H5)3
CAS Number:
Molecular Weight:
190.31
Beilstein:
1767229
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

≥98%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

160-161 °C (lit.)
62-63 °C/20 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)C=C

InChI

1S/C8H18O3Si/c1-5-9-12(8-4,10-6-2)11-7-3/h8H,4-7H2,1-3H3

InChI key

FWDBOZPQNFPOLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geun Yeol Bae et al.
Journal of colloid and interface science, 344(2), 584-587 (2010-02-09)
Superhydrophobic poly(lactic acid) (PLA) fabrics are prepared by UV photo-grafting of hydrophobic silica particles possessing vinyl functional groups on the surfaces, which is a novel one-step process to provide surface with roughness as well as hydrophobicity simultaneously. For this purpose
Bindu Prasannakumaran Nair et al.
Langmuir : the ACS journal of surfaces and colloids, 26(2), 730-735 (2009-10-09)
Facile, surfactant free synthetic strategy for bifunctionalized hybrid silica spheres (HS) with structural ordering is presented. HS was prepared by casting and drying of stable siloxane solution from hydrolytic co-condensation of 3-aminopropyltriethoxysilane (AS) and vinyltriethoxysilane (VS) with AS:VS mole ratio
David Tulumello et al.
The journal of physical chemistry. B, 109(13), 6343-6354 (2006-07-21)
The C 1s, Si 2p, Si 2s, and O 1s inner-shell excitation spectra of vinyltriethoxysilane, trimethylethoxysilane, and vinyltriacetoxysilane have been recorded by electron energy loss spectroscopy under scattering conditions dominated by electric dipole transitions. The spectra are converted to absolute
Ying-Sing Li et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(12), 2759-2766 (2004-09-08)
Infrared and Raman spectra of vinyltriethoxysilane (VTES), the VTES sol-gel, and the sol-gel coated aluminum have been collected. The assignments of the vibrational modes for the silane, the sol-gel, and the sol-gel films have been made based on the group
S Fink
Stain technology, 62(1), 27-33 (1987-01-01)
As a new aqueous adhesive to affix sections to glass slides, hydrolyzed vinyltriethoxysilane--either pure, in combination with polyvinyl alcohol or with specially prepared aqueous polyacrylate solutions--was applied. The silane proved to be very effective in enhancing bonding to the glass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service