Skip to Content
Merck
All Photos(1)

Key Documents

672033

Sigma-Aldrich

(S)-N-Boc-2,3-epoxypropylamine

97%

Synonym(s):

tert-Butyl (2S)-N-(2-oxiranylmethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.5% (GCF)
97%

form

solid

optical purity

ee: ≥99.0% (GC)

mp

48-50 °C

functional group

amine
ether

SMILES string

CC(C)(C)OC(=O)NC[C@H]1CO1

InChI

1S/C8H15NO3/c1-8(2,3)12-7(10)9-4-6-5-11-6/h6H,4-5H2,1-3H3,(H,9,10)/t6-/m0/s1

InChI key

ZBBGKXNNTNBRBH-LURJTMIESA-N

Application

(S)-N-Boc-2,3-epoxypropylamine can be used:
  • To functionalize detonation nanodiamonds with NH2 functional group to enhance their optical properties.
  • In the synthesis of poly(ethylene glycol) (PEG) based block copolymers, which are further used in the preparation of micelles loaded with mRNA for intracellular mRNA delivery.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanidine-phosphate interactions stabilize polyion complex micelles based on flexible catiomers to improve mRNA delivery
Miyazaki T, et al.
European Polymer Journal, 140(1), 110028-110028 (2020)
Tandem malonate-based glucosides (TMGs) for membrane protein structural studies
Hussain H, et al.
Scientific reports, 7(1), 1-11 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service