Skip to Content
Merck
All Photos(1)

Key Documents

671525

Sigma-Aldrich

4,7,10,13,16,19,22,25,32,35,38,41,44,47,50,53-Hexadecaoxa-28,29-dithiahexapentacontanedioic acid

≥95%

Synonym(s):

PEG acid disulfide (n=7), PEG propionate disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H74O20S2
CAS Number:
Molecular Weight:
915.11
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (oligomer purity)
≥95%

form

solid

reaction suitability

reaction type: Pegylations

storage temp.

−20°C

SMILES string

OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCSSCCOCCOCCOCCOCCOCCOCCOCCOCCC(O)=O

InChI

1S/C38H74O20S2/c39-37(40)1-3-43-5-7-45-9-11-47-13-15-49-17-19-51-21-23-53-25-27-55-29-31-57-33-35-59-60-36-34-58-32-30-56-28-26-54-24-22-52-20-18-50-16-14-48-12-10-46-8-6-44-4-2-38(41)42/h1-36H2,(H,39,40)(H,41,42)

InChI key

SSUZECNFRDBXHC-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of orthogonally dual-clickable Janus nanoparticles via cyclic templating strategy
  • Preparation of cyclic RGD functionalized gold nanoparticles for tumor targeting
  • Asymmetric-functionalized nanoparticle with orthogonal clickable functionalities
  • Functional immobilization of small GTPase Rab6A on DNA-gold nanoparticles by using a site-specifically attached poly(ethylene glycol) linker and thiol-place exchange reaction
  • Direct writing of metal nanoparticle films inside sealed microfluidic channels in relation to biofunctionalization with ligands for protein capture and fluorescence assay

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service