Skip to Content
Merck
All Photos(1)

Key Documents

670820

Sigma-Aldrich

Pam3-Cys-OH

≥98.0% (TLC), for peptide synthesis

Synonym(s):

N-α-Palmitoyl-S-[2,3-bis(palmitoyloxy)-(2RS)-propyl]-L-cysteine, Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H103NO7S
CAS Number:
Molecular Weight:
910.46
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Product Name

Pam3-Cys-OH, ≥98.0% (TLC)

Assay

≥98.0% (TLC)

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CCCCCCCCCCCCCCCC(=O)N[C@@H](CSCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC)C(O)=O

InChI

1S/C54H103NO7S/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-51(56)55-50(54(59)60)48-63-47-49(62-53(58)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)46-61-52(57)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h49-50H,4-48H2,1-3H3,(H,55,56)(H,59,60)/t49?,50-/m0/s1

InChI key

PZFZLRNAOHUQPH-GOOVXGPGSA-N

General description

Pam3-Cys-OH is a lipopeptide containing a cysteine residue at the C-terminus, often used in solid phase peptide synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipoconjugates for the noncovalent generation of microarrays in biochemical and cellular assays
A Hoff, et al.
Chembiochem, 3, 1183-1191 (2002)
Katharina A Remer et al.
Journal of immunological methods, 313(1-2), 1-10 (2006-05-25)
THP-1 2A9, a subclone of the monocytoid cell line THP-1 and known to be exquisitely sensitive to LPS, was tested for TNF production following triggering by excess doses of TLR ligands. TLR2, TLR4 and TLR5 agonists, but neither TLR3 nor
Amrita D Joshi et al.
Journal of leukocyte biology, 83(2), 314-324 (2007-11-22)
Macrophages play a pivotal role in innate and acquired immune responses to Schistosoma mansoni. Classical (M1) or alternative (M2) activation states of these cells further delineate their roles in tissue damage through innate immunity or fibrotic remodeling, respectively. In the
Sudha Natarajan et al.
Shock (Augusta, Ga.), 33(2), 162-169 (2009-06-03)
LPS challenge causes potent activation of innate immunity. Because LPS is ubiquitously present in ambient air, repeated inhalation may lead to activation of the pulmonary immune response. If this activation is unregulated, chronic LPS inhalation would lead to persistent inflammation
Pitchaimani Kandasamy et al.
The Journal of biological chemistry, 286(10), 7841-7853 (2011-01-06)
Mycoplasma pneumoniae is a human pathogen causing respiratory infections that are also associated with serious exacerbations of chronic lung diseases. Membranes and lipoproteins from M. pneumoniae induced a 4-fold increase in arachidonic acid (AA) release from RAW264.7 and a 2-fold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service