Skip to Content
Merck
All Photos(1)

Key Documents

667897

Sigma-Aldrich

1,2-Diethoxy-1,1,2,2-tetramethyldisilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3CH2O)(CH3)2Si)2
CAS Number:
Molecular Weight:
206.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.423

bp

84 °C/50 mmHg

density

0.836 g/mL at 25 °C

SMILES string

CCO[Si](C)(C)[Si](C)(C)OCC

InChI

1S/C8H22O2Si2/c1-7-9-11(3,4)12(5,6)10-8-2/h7-8H2,1-6H3

InChI key

GWIVSKPSMYHUAK-UHFFFAOYSA-N

Application

Disilane reagent used to prepare dimethylsilanol cross-coupling partners.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F

Flash Point(C)

42 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott E Denmark et al.
Organic letters, 5(19), 3483-3486 (2003-09-12)
[reaction: see text] A mild and general palladium-catalyzed insertion of 1,2-diethoxy-1,1,2,2-tetramethyldisilane into a variety of aryl bromides affords the aryldimethylsilyl ethers in high yields. Hydrolysis of the ethers under pH-optimized conditions results in the exclusive formation of the desired aryldimethylsilanols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service