Skip to Content
Merck
All Photos(3)

Documents

667609

Sigma-Aldrich

Bis[2-(N,N-dimethylamino)ethyl] ether

97%

Synonym(s):

2,2′-Oxybis(N,N-dimethylethylamine), 2-(Dimethylamino)ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)2N(CH2CH2))2O
CAS Number:
Molecular Weight:
160.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.430

bp

189 °C/760 mmHg

density

0.841 g/mL at 25 °C

SMILES string

CN(C)CCOCCN(C)C

InChI

1S/C8H20N2O/c1-9(2)5-7-11-8-6-10(3)4/h5-8H2,1-4H3

InChI key

GTEXIOINCJRBIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Dimethylamino)ethyl ether is a tridentate ligand. It assists in the formation and stabilization of intermediates during the exchange reactions.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

151.0 °F

Flash Point(C)

66.11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ballantyne
Veterinary and human toxicology, 39(5), 290-295 (1997-10-06)
Bis[2-(dimethylamino)ethyl]ether (DMAEE; CAS No 3033-62-3) is an industrial liquid chemical used principally as an amine catalyst. It was investigated for potential acute hazards. DMAEE is of moderate acute peroral toxicity with LD50 values in the rat of 1045 ml/kg and
Bis[2-(dimethylamino)ethyl]ether(CH3)2NCH2CH2OCH2CH2N(CH3)2.
B Ballantyne
Journal of applied toxicology : JAT, 25(3), 260-269 (2005-04-28)
Chao-Shan Da et al.
Organic letters, 11(24), 5578-5581 (2009-11-17)
Generally used and highly reactive RMgBr reagents were effectively deactivated by bis[2-(N,N-dimethylamino)ethyl] ether and then were employed in the highly enantioselective addition of Grignard reagents to aldehydes. The reaction was catalyzed by the complex of commercially available (S)-BINOL and Ti(O(i-)Pr)(4)
Mild Iodine- Magnesium Exchange of Iodoaromatics Bearing a Pyrimidine Ring with Isopropylmagnesium Chloride
Wang XJ, et al.
Organic Letters, 8, 3141-3144 (2006)
Se-Ra Shin et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-10)
Isosorbide (ISB), a nontoxic bio-based bicyclic diol composed from two fuzed furans, was incorporated into the preparation of flexible polyurethane foams (FPUFs) for use as a cell opener and to impart antioxidant properties to the resulting foam. A novel method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service