Skip to Content
Merck
All Photos(1)

Key Documents

659967

Sigma-Aldrich

Heterogeneous Palladium Catalysts Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

Quality Level

General description

It contains a set of six different heterogeneous palladium catalysts, useful for rapid screening of catalysis conditions. It is in sampler format with individual components packaged for multiple experiments and mini scale-up. The cost of the kit is less than the total cost of individual components.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 205699Palladium on carbon, extent of labeling: 10 wt. % loading, matrix activated carbon support 1 gSDS

  • 205737Palladium on calcium carbonate, extent of labeling: 5 wt. % loading, poisoned with lead 1 gSDS

  • 212911Palladium hydroxide on carbon, extent of labeling: 20 wt. % loading (dry basis), matrix carbon, wet support 1 gSDS

  • 277991Palladium on barium sulfate, reduced, extent of labeling: 5 wt. % loading 1 gSDS

  • 330108Palladium on carbon, Evonik NOBLYST® P1173 10% Pd 1 gSDS

  • 440086Palladium on alumina, extent of labeling: 10 wt. % loading, powder, reduced, dry 1 gSDS

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 1 - Eye Irrit. 2 - Lact. - Repr. 1A - Skin Irrit. 2 - STOT RE 2

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service