656321
3-Chloro-4-methoxyphenethylamine hydrochloride
90%
Synonym(s):
2-(3-Chloro-4-methoxyphenyl)ethylamine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12ClNO · HCl
CAS Number:
Molecular Weight:
222.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
90%
form
solid
functional group
amine
chloro
SMILES string
Cl.COc1ccc(CCN)cc1Cl
InChI
1S/C9H12ClNO.ClH/c1-12-9-3-2-7(4-5-11)6-8(9)10;/h2-3,6H,4-5,11H2,1H3;1H
InChI key
VGUBBPZQDURYFN-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service