650897
(4S,4′S)-(−)-2,2′-(3-Pentylidene)bis(4-isopropyloxazoline)
97%
Synonym(s):
(4S,4′S)-(−)-2,2′-(1-Ethylpropylidene)bis(4,5-dihydro-4-isopropyloxazole), [S-(R*,R*)]-(−)-2,2′-(1-Ethylpropylidene)bis(4,5-dihydro-4-isopropyloxazole)
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.4710 (lit.)
bp
120 °C/0.001 mmHg (lit.)
density
1.002 g/mL at 25 °C (lit.)
functional group
ether
SMILES string
CCC(CC)(C1=N[C@H](CO1)C(C)C)C2=N[C@H](CO2)C(C)C
InChI
1S/C17H30N2O2/c1-7-17(8-2,15-18-13(9-20-15)11(3)4)16-19-14(10-21-16)12(5)6/h11-14H,7-10H2,1-6H3/t13-,14-/m1/s1
InChI key
PIJFQTUSLZLNOP-ZIAGYGMSSA-N
Related Categories
Application
- To prepare [11C]carboxylic acids, [11C]esters and [11C]amides using boronic acid esters and 11CO2.
- In one of the key synthetic steps for the total synthesis of tatanan A and 3-epi-tatanan A.
- To prepare copper(I) complex and used in a mass spectrometric method to study its chiral selectivity with different substrates.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
180.0 °F - closed cup
Flash Point(C)
82.22 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service