Skip to Content
Merck
All Photos(1)

Key Documents

645168

Sigma-Aldrich

Tungsten(VI) chloride

powder, ≥99.99% trace metals basis (purity excludes molybdenum)

Synonym(s):

Tungsten hexachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WCl6
CAS Number:
Molecular Weight:
396.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis (purity excludes molybdenum)
form:
powder
Pricing and availability is not currently available.

vapor pressure

43 mmHg ( 215 °C)

Quality Level

Assay

≥99.99% trace metals basis (purity excludes molybdenum)

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

bp

347 °C (lit.)

mp

275 °C (lit.)

density

3.52 g/mL at 25 °C (lit.)

SMILES string

Cl[W](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.W/h6*1H;/q;;;;;;+6/p-6

InChI key

KPGXUAIFQMJJFB-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chongshen Guo et al.
Biomaterials, 52, 407-416 (2015-03-31)
Photothermal ablation (PTA) therapy has been viewed as an invasive option for cancer therapy with minimal deconstruction of healthy tissues. In this study, a potent candidate of (NH4)xWO3 nanocube was developed for PTA treatment of metastatic breast cancer in the
Tobias Fält et al.
Journal of computer assisted tomography, 39(6), 975-980 (2015-08-22)
The aim of this study was to determine the potential of different high-Z elements to act as contrast media (CMs) alongside iodine (I) in dual-CM, dual-energy (DE) computed tomography examinations. Gadolinium (Gd), tantalum (Ta), wolfram (W), gold (Au), and bismuth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service