Skip to Content
Merck
All Photos(1)

Key Documents

632732

Sigma-Aldrich

1-Boc-pyrazole-4-boronic acid pinacol ester

97%

Synonym(s):

1,1-dimethylethyl ester 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Pyrazole-1-carboxylic acid, 1-tert-Butoxycarbonyl-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)pyrazole, 1-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-Boc-4-pyrazoleboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-Boc-pyrazole, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylic acid tert-butyl ester, tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-pyrazolecarboxylate, [1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl]boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23BN2O4
Molecular Weight:
294.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

82-86 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)n1cc(cn1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C14H23BN2O4/c1-12(2,3)19-11(18)17-9-10(8-16-17)15-20-13(4,5)14(6,7)21-15/h8-9H,1-7H3

InChI key

IPISOFJLWYBCAV-UHFFFAOYSA-N

Application

Reagent used for
  • Suzuki Coupling
  • Copper-catalyzed azidation

Reagent used in Preparation of
  • Selective quinazolinyl-phenol inhibitors of CHK1 as potential antitumors and radioprotectants
  • Stereoselective synthesis of selective Cathepsin inhibitors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(II)-catalyzed conversion of aryl/heteroaryl boronic acids, boronates, and trifluoroborates into the corresponding azides: substrate scope and limitations
K. D. Grimes, et al.,
Synthesis, 9, 1441-1448 (2010)
John J Caldwell et al.
Journal of medicinal chemistry, 54(2), 580-590 (2010-12-29)
Structure-based design was applied to the optimization of a series of 2-(quinazolin-2-yl)phenols to generate potent and selective ATP-competitive inhibitors of the DNA damage response signaling enzyme checkpoint kinase 2 (CHK2). Structure-activity relationships for multiple substituent positions were optimized separately and
Paul A Bethel et al.
Bioorganic & medicinal chemistry letters, 19(16), 4622-4625 (2009-07-21)
A number of molecular recognition features have been exploited in structure-based design of selective Cathepsin inhibitors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service