Skip to Content
Merck
All Photos(3)

Key Documents

631493

Sigma-Aldrich

Tricyclohexylphosphine tetrafluoroborate

97%

Synonym(s):

Tricyclohexylphosphonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)3P · HBF4
CAS Number:
Molecular Weight:
368.24
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Arylations
reaction type: Cross Couplings
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

mp

164 °C (lit.)

functional group

phosphine

SMILES string

F[B-](F)(F)F.[H][P+](C1CCCCC1)(C2CCCCC2)C3CCCCC3

InChI

1S/C18H33P.BF4/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1(3,4)5/h16-18H,1-15H2;/q;-1/p+1

InChI key

MYSMMEUXKHJYKH-UHFFFAOYSA-O

General description

Tricyclohexylphosphine tetrafluoroborate (PCy3·HBF4) is an inexpensive and air-stable phosphine ligand, commonly used in cross-coupling reactions.

Application

Tricyclohexylphosphine tetrafluoroborate may be used in the following processes:
  • As a ligand for preparing C-homoaporphine alkaloids via microwave-assisted direct-arylation.
  • To synthesize poly-[9,9-bis(3-propylamide-2-methylpropyl sulfonic acid) fluorene]-co-(4,4′-diphenyl) (PFDBSO3H), which can be employed as a template and doping agent for enhancing the conductivity of poly(3,4-ethylenedioxythiophene) (PEDOT) films.
  • To improve the reactivity of palladium-catalyzed Suzuki-Miyaura cross-coupling reaction between MIDA boronates and less activated alkenyl tosylates.
Used with Ru(cod)Cl dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of PEDOT aqueous dispersions with sulfonated polyfluorene as a template and doping agent.
Gu J, et al.
Reactive functional Polymers, 100, 83-88 (2016)
Synthesis of C-homoaporphines via microwave-assisted direct arylation.
Chaudhary S and Harding WW.
Tetrahedron, 67(3), 569-575 (2011)
Suzuki-Miyaura cross-coupling of alkenyl tosylates with alkenyl MIDA boronates.
Luthy M and Taylor RJK.
Tetrahedron Letters, 53(27), 3444-3447 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service