All Photos(1)
About This Item
Linear Formula:
ClC6H4≡CH
CAS Number:
Molecular Weight:
136.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.5640 (lit.)
bp
178-179 °C (lit.)
density
1.109 g/mL at 25 °C (lit.)
functional group
chloro
SMILES string
Clc1cccc(c1)C#C
InChI
1S/C8H5Cl/c1-2-7-4-3-5-8(9)6-7/h1,3-6H
InChI key
GRBJPHPMYOUMJV-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
145.0 °F - closed cup
Flash Point(C)
62.78 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service