61630
Ethyl dodecanoate
≥98.0% (GC)
Synonym(s):
Ethyl laurate, Lauric acid ethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)10COOC2H5
CAS Number:
Molecular Weight:
228.37
Beilstein:
1769671
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0% (GC)
bp
269 °C (lit.)
density
0.862 g/mL at 20 °C (lit.)
functional group
ester
SMILES string
CCCCCCCCCCCC(=O)OCC
InChI
1S/C14H28O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h3-13H2,1-2H3
InChI key
MMXKVMNBHPAILY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D R Bevan et al.
Cancer letters, 57(2), 173-179 (1991-05-01)
Effects of vehicles and of asbestos on disposition of benzo[a]pyrene (B[a]P) were examined in vivo. [3H]B[a]P dissolved in triethylene glycol, ethyl laurate, or tricaprylin was intratracheally administered to male Sprague-Dawley rats. Excretion was most rapid when triethylene glycol was the
Development of an ethyl laurate-based microemulsion for rapid-onset intranasal delivery of diazepam.
Lianli Li et al.
International journal of pharmaceutics, 237(1-2), 77-85 (2002-04-17)
An ethyl laurate-based microemulsion system with Tween 80 as surfactant, propylene glycol and ethanol as cosolvents was developed for intranasal delivery of diazepam. Phase behavior and solubilization capacity of the microemulsion system were characterized and in vivo nasal absorption of
A Botsi et al.
Carbohydrate research, 241, 37-46 (1993-03-17)
The inclusion complexes of cyclomaltoheptaose (beta CD) and heptakis(2,3,6-tri-O-methyl)cyclomaltoheptaose (TM-beta CD) with the four major components of the pheromone of the olive fruit fly (Dacus oleae), namely 1,7-dioxaspiro[5.5]undecane, (-)-alpha-pinene, nonanal, and ethyl dodecanoate, and the complex of heptakis(2,6-di-O-methyl)cyclomaltoheptaose (DM-beta CD)
Hydrocracking of ethyl laurate on bifunctional micro-/mesoporous zeolite catalysts.
Oliver Busse et al.
ChemSusChem, 3(5), 563-565 (2010-05-04)
Napaporn Komesvarakul et al.
Journal of cosmetic science, 57(4), 309-325 (2006-09-08)
Microemulsification of triglyceride-based oil is challenging due to the formation of undesirable phases such as macroemulsions, liquid crystals, or sponge phases. This research evaluates the formation of artificial sebum microemulsions using linker molecules, with the addition of co-oil to help
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service