Skip to Content
Merck
All Photos(1)

Key Documents

615439

Sigma-Aldrich

Methyl-d3-triethoxysilane

99 atom % D, 97% (CP)

Synonym(s):

Triethoxy(methyl-d3)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3Si(OC2H5)3
Molecular Weight:
181.32
UNSPSC Code:
12352005

isotopic purity

99 atom % D

Assay

97% (CP)

bp

141-143 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)C([2H])([2H])[2H]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service