597449
3-Thiophenecarbonitrile
95%
Synonym(s):
3-Cyanothiophene, Thiophene-3-nitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H3NS
CAS Number:
Molecular Weight:
109.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
refractive index
n20/D 1.5630 (lit.)
bp
82 °C/10 mmHg (lit.)
density
1.2 g/mL at 25 °C (lit.)
functional group
nitrile
SMILES string
N#Cc1ccsc1
InChI
1S/C5H3NS/c6-3-5-1-2-7-4-5/h1-2,4H
InChI key
GSXCEVHRIVLFJV-UHFFFAOYSA-N
Application
3-Thiophenecarbonitrile can be used to synthesize the following:
- 2,4-dinaphthyl-3-cyanothiophene via reaction with iodonaphthalene
- 2,5-bis-tri(n-butyl)tin-3-cyanothiophene, a building block for the synthesis of new alternating (3-alkyl/3-cyano)thiophene copolymers
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
186.0 °F - closed cup
Flash Point(C)
85.56 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
"Extension of the Heck reaction to the arylation of activated thiophenes"
<BIG>Lavenot.L, et al.</BIG>
Journal of Organometallic Chemistry, 567(01), 49-55 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service