Skip to Content
Merck
All Photos(1)

Key Documents

588350

Sigma-Aldrich

Bromomethane-d2

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CD2HBr
CAS Number:
Molecular Weight:
96.95
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

bp

4 °C (lit.)

application(s)

environmental

mass shift

M+2

SMILES string

[2H]C([2H])Br

InChI

1S/CH3Br/c1-2/h1H3/i1D2

InChI key

GZUXJHMPEANEGY-DICFDUPASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Muta. 2 - Ozone 1 - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service